missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Human IgG Fc Recombinant Secondary Antibody, HRP

Product Code. 17858791
Change view
Click to view available options
Quantity:
50 μL
250 μL
Unit Size:
250µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17858791 250 μL 250µL
17818801 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17858791 Supplier Invitrogen™ Supplier No. A55745
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Goat Recombinant Monoclonal Secondary Antibody

This Goat anti-Human IgG Fc Recombinant Secondary Antibody, HRP specifically detects human IgG Fc with no cross reactivity to human IgA or IgM or various species immunoglobulins. The sensitivity and specificity of each lot is confirmed using ELISA. Recombinant antibodies are produced using specific genes that code for the desired antibodies. These genes are cloned into an expression vector and expressed in vitro. The advantages of recombinant antibodies include: better specificity, animal origin-free formulation, and lot-to-lot consistency. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG Fc
Applications Peptide Array, ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3H8L9
Concentration 1.0 mg/mL
Conjugate HRP
Formulation PBS with proprietary stabilizer and proprietary preservative; pH 7.2-7.4
Host Species Goat
Immunogen Purified Human IgG.
Purification Method Protein G
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.