Learn More
Invitrogen™ Goat anti-Human IgE Cross-Adsorbed Secondary Antibody, FITC
Goat Polyclonal Secondary Antibody
Brand:  Invitrogen™ A18800
Product Code. 15321246
  • £321.00 / 1mg
Description
Goat anti-Human IgE Cross-Adsorbed Secondary Antibody for Flow, ICC/IF
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Human IgE Cross-Adsorbed
Polyclonal
FITC
Goat
Affinity chromatography
RUO
Human
Secondary Antibody
IgG
Flow Cytometry, Immunocytochemistry
1.5 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.8
Human IgE.
1 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.