missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgD Heavy Chain Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68528V
This item is not returnable.
View return policy
Description
IgD Heavy Chain Polyclonal antibody specifically detects IgD Heavy Chain in Human samples. It is validated for ELISA
Specifications
| IgD Heavy Chain | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
| ELISA | |
| DyLight 405 | |
| Constant Region of Heavy Chain of IgD, Ig Delta Chain C Region, Ig Delta Chain C Region NIG-65, Ig Delta Chain C Region WAH, Immunoglobulin Delta, immunoglobulin heavy constant delta | |
| Human IgD paraproteins | |
| 1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction