missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA Heavy Chain Secondary Antibody, PerCP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68301PCP
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| PBS | |
| Highly purified human IgA alpha-chain | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| PerCP | |
| Goat | |
| Affinity purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction