missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgA Heavy Chain Secondary Antibody, DyLight 550 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-75759
242.25 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
IgA Heavy Chain Polyclonal antibody specifically detects IgA Heavy Chain in Human samples. It is validated for Flow Cytometry, Immunomicroscopy.
Specifications
| IgA Heavy Chain | |
| Polyclonal | |
| DyLight 550 | |
| Immunoglobulin A | |
| Goat | |
| Affinity purified | |
| RUO | |
| 3493 | |
| Rehydrate with 1.1 ml of deionized water and let stand 30 minutes at room temperature to dissolve. (Product has been overfilled to ensure complete recovery.) Centrifuge to remove any particulates. Prepare fresh working dilution daily. | |
| Store lyophilized antibody at 4°C. Mix reconstituted liquid with an equal volume of glycerol, aliquot contents and freeze at -20°C or below for long term storage. |
| Flow Cytometry, Immunomicroscopy | |
| LYOPH | |
| Flow Cytometry 1:20 - 1:2000, Immunomicroscopy 1:20 - 1:2000 | |
| IGHA1 | |
| Purified human IgA, (a chain) | |
| 1 mg | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy (alpha) chains on human IgA | |
| Human | |
| IgA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction