Learn More
HNF1A, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of HNF1A.
Brand: Abnova PAB7190.100ug
Description
The protein encoded by this gene is a transcription factor required for the expression of several liver-specific genes. The encoded protein functions as a homodimer and binds to the inverted palindrome 5'-GTTAATNATTAAC-3'. Defects in this gene are a cause of maturity onset diabetes of the young type 3 (MODY3) and also can result in the appearance of hepatic adenomas. [provided by RefSeq]
Sequence: C-DELPTKKGRRNRFKSpecifications
| HNF1A | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) The optimal working dilution should be determined by the end user. | |
| HNF1A | |
| HNF1A | |
| A synthetic peptide corresponding to human HNF1A. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of HNF1A. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| HNF1/LFB1/MODY3/TCF1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 6927 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only