Learn More
Invitrogen™ Goat anti-Syrian/Armenian Hamster IgG (H+L) Secondary Antibody, Biotin
Description
Product # 31750 has been successfully used in Western blot, IF, ICC, IHC, ELISA and ISH applications. Product # 31750 reacts with the heavy chains of syrian hamster IgG and with the light chains common to most hamster immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody cross-reacts with horse IgG, human IgG, rat IgG, and swine IgG (3%), bovine IgG, cat IgG, dog IgG, mouse IgG, rat IgM, and Rhesus monkey IgG (1%) and it has minimum cross-reactivity with immunoglobulins from other species. This anti-hamster IgG antibody conjugate recognizes IgG from both Syrian and Armenian hamsters.
Specifications
Specifications
| Antigen | Syrian/Armenian Hamster IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 3mg/mL BSA and 0.08% sodium azide; pH 7.8 |
| Host Species | Goat |
| Purification Method | Affinity chromatography |
| Quantity | 1.5 mg |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.