missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Hamster IgG (H+L) Secondary Antibody, Allophycocyanin/Cy7 (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-71730APCCY7
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Dot Blot, Fluorophore-linked immunosorbent assay
Specifications
| IgG (H+L) | |
| Polyclonal | |
| PBS | |
| Armenian and Golden Syrian Hamster IgG, whole molecule | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Hamster | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Dot Blot | |
| APC-Cyanine7 | |
| Goat | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction