missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Guinea pig IgG whole molecule, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Brand: Abnova PAB10614.2mg
This item is not returnable.
View return policy
Specifications
| Goat anti-Guinea pig IgG whole molecule | |
| Polyclonal | |
| Goat anti-guinea pig IgG recognizes Guinea pig IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Guinea pig IgG whole molecule | |
| 2 mg | |
| Guinea Pig | |
| Liquid |
| Flow Cytometry, Immunofluorescence | |
| Texas Red | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction