missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Guinea Pig IgG Fc Secondary Antibody, DyLight 405, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60685V
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Guinea Pig samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
| IgG Fc | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Guinea Pig IgG F(c) fragment | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Guinea Pig | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| DyLight 405 | |
| Goat | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction