Learn More
GRIN1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GRIN1.
Brand: Abnova PAB7074.100ug
Description
The protein encoded by this gene is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described. [provided by RefSeq
Sequence: C-TQERVNNSNKKESpecifications
| GRIN1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| GRIN1 | |
| NMDA1/NMDAR1/NR1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 2902 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of GRIN1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000823.4;NP_067544.1;NP_015566.1 | |
| GRIN1 | |
| A synthetic peptide corresponding to human GRIN1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize all three reported isoforms (NP_000823.4, NP_067544.1, NP_015566.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only