Learn More
GPX6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GPX6.
Brand: Abnova PAB7066.100ug
Description
This gene product belongs to the glutathione peroxidase family, which functions in the detoxification of hydrogen peroxide. It contains a selenocysteine (Sec) residue at its active site. The selenocysteine is encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Expression of this gene is restricted to embryos and adult olfactory epithelium. [provided by RefSeq]
Sequence: QTLKPQNRKVDCSpecifications
| GPX6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) The optimal working dilution should be determined by the end user. | |
| GPX6 | |
| GPX6 | |
| A synthetic peptide corresponding to human GPX6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of GPX6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_874360.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 257202 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only