missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR83 Goat anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GPR83.
Brand: Abnova PAB24579.100ug
This item is not returnable.
View return policy
Description
Sequence: C-RRYGAESQNPTVKSpecifications
| GPR83 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of GPR83. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_057624.3 | |
| GPR83 | |
| A synthetic peptide corresponding to amino acids 59-71 at internal region of human GPR83. | |
| 100 μg | |
| Primary | |
| The immunizing peptide represents part of an extracellular domain. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR83 | |
| GIR/GPR72 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10888 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction