missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GOLGA6A/GOLGA6B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GOLGA6A/GOLGA6B.
Brand: Abnova PAB6865.100ug
This item is not returnable.
View return policy
Description
Sequence: C-DLPKEKADGTEQSpecifications
| GOLGA6A/GOLGA6B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| NP_0010.33729.1 (Gene ID : 342096);NP_061122.4 (Gene ID : 55889) | |
| GOLGA6A | |
| A synthetic peptide corresponding to internal region of human GOLGA6A/GOLGA6B. | |
| 100 μg | |
| Primary | |
| GeneID 55889 may be a pseudogene, while GeneID 342096 is thought to encode a functional protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of GOLGA6A/GOLGA6B. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| FLJ75859/GLP | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 342096/55889 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction