Learn More
GM2A Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GM2A.
Brand: Abnova PAB19672.100ug
Description
This gene encodes a small glycolipid transport protein which acts as a substrate specific co-factor for the lysosomal enzyme beta-hexosaminidase A. Beta-hexosaminidase A, together with GM2 ganglioside activator, catalyzes the degradation of the ganglioside GM2, and other molecules containing terminal N-acetyl hexosamines. Mutations in this gene result in GM2-gangliosidosis type AB or the AB variant of Tay-Sachs disease. [provided by RefSeq
Sequence: KRLGCIKIAASSpecifications
| GM2A | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of GM2A. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000396.2 | |
| GM2A | |
| A synthetic peptide corresponding to amino acids 179-189 at internal region of human GM2A. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_000396.2) only. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| GM2A | |
| SAP-3 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 2760 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only