Learn More
GJB6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GJB6.
Brand: Abnova PAB17269.100ug
Description
Gap junctions allow the transport of ions and metabolites between the cytoplasm of adjacent cells. They are formed by two hemichannels, made up of six connexin proteins assembled in groups. Each connexin protein has four transmembrane segments, two extracellular loops, a cytoplasmic loop formed between the two inner transmembrane segments, and the N- and C-terminus both being in the cytoplasm. The specificity of the gap junction is determined by which connexin proteins comprise the hemichannel. In the past, connexin protein names were based on their molecular weight, however the new nomenclature uses sequential numbers based on which form (alpha or beta) of the gap junction is present. This gene encodes one of the connexin proteins. Mutations in this gene have been found in some forms of deafness and in some families with hidrotic ectodermal dysplasia. [provided by RefSeq
Sequence: C-RSKRAQTQKNHPNHSpecifications
| GJB6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| GJB6 | |
| CX30/DFNA3/ED2/EDH/HED | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10804 | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of GJB6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_006774.2 | |
| GJB6 | |
| A synthetic peptide corresponding to amino acids at internal region of human GJB6. | |
| 100 μg | |
| Primary | |
| Reported variants represent identical protein: NP_006774.2, NP_001103691.1, NP_001103689.1, NP_001103690.1 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only