Learn More
GCNT3 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GCNT3.
Brand: Abnova PAB7556.100ug
Description
This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins
Sequence: C-NKFDPKVDDNALQSpecifications
| GCNT3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| GCNT3 | |
| C2/4GnT/C2GnT-M/C2GnT2/GnT-M | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9245 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of GCNT3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_004742.1 | |
| GCNT3 | |
| A synthetic peptide corresponding to amino acids 410-422 of human GCNT3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only