Learn More
GALR1 Goat anti-Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of GALR1.
Brand: Abnova PAB7173.100ug
Description
The neuropeptide galanin elicits a range of biological effects by interaction with specific G-protein-coupled receptors. Galanin receptors are seven-transmembrane proteins shown to activate a variety of intracellular second-messenger pathways. GALR1 inhibits adenylyl cyclase via a G protein of the Gi/Go family. GALR1 is widely expressed in the brain and spinal cord, as well as in peripheral sites such as the small intestine and heart. [provided by RefSeq
Sequence: C-RSKPGKPRSTTNSpecifications
| GALR1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| GALR1 | |
| GALNR/GALNR1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 2587 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of GALR1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001471.1 | |
| GALR1 | |
| A synthetic peptide corresponding to human GALR1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only