missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FTCD Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FTCD.
Brand: Abnova PAB6664.100ug
This item is not returnable.
View return policy
Description
FTCD is a bifunctional enzyme that channels 1-carbon units from formiminoglutamate, a metabolite of the histidine degradation pathway, to the folate pool.[supplied by OMIM
Sequence: SQLVECVPNFSEGKNQSpecifications
| FTCD | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western blot (0.03-0.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| FTCD | |
| LCHC1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 10841 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FTCD. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_006648;NP_996848 | |
| FTCD | |
| A synthetic peptide corresponding to human FTCD. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (as represented by NP_006648 and NP_996848). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction