missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXN2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FOXN2.
Brand: Abnova PAB6445.100ug
This item is not returnable.
View return policy
Description
This gene encodes a forkhead domain binding protein and may function in the transcriptional regulation of the human T-cell leukemia virus long terminal repeat. [provided by RefSeq
Sequence: MCDPLDQLATRTQKSpecifications
| FOXN2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| FOXN2 | |
| HTLF | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 3344 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FOXN2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_002149 | |
| FOXN2 | |
| A synthetic peptide corresponding to human FOXN2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction