Learn More
FGFR1 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FGFR1.
Brand: Abnova PAB6742.100ug
Description
The protein encoded by this gene is a member of the fibroblast growth factor receptor (FGFR) family, where amino acid sequence is highly conserved between members and throughout evolution. FGFR family members differ from one another in their ligand affinities and tissue distribution. A full-length representative protein consists of an extracellular region, composed of three immunoglobulin-like domains, a single hydrophobic membrane-spanning segment and a cytoplasmic tyrosine kinase domain. The extracellular portion of the protein interacts with fibroblast growth factors, setting in motion a cascade of downstream signals, ultimately influencing mitogenesis and differentiation. This particular family member binds both acidic and basic fibroblast growth factors and is involved in limb induction. Mutations in this gene have been associated with Pfeiffer syndrome, Jackson-Weiss syndrome, Antley-Bixler syndrome, osteoglophonic dysplasia, and autosomal dominant Kallmann syndrome 2. Chromosomal aberrations involving this gene are associated with stem cell myeloproliferative disorder and stem cell leukemia lymphoma syndrome. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq
Sequence: CLPRHPAQLANGGLKRSpecifications
| FGFR1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western blot (0.3-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| FGFR1 | |
| BFGFR/CD331/CEK/FGFBR/FLG/FLJ99988/FLT2/HBGFR/KAL2/N-SAM | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 2260 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FGFR1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000595;NP_056934;NP_075593;NP_075594;NP_075599 | |
| FGFR1 | |
| A synthetic peptide corresponding to human FGFR1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize 5 (out of nine reported) isoforms (as represented by NP_000595, NP_056934, NP_075593, NP_075594, NP_075599). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only