Learn More
FEZ1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FEZ1.
Brand: Abnova PAB7151.100ug
Description
This gene is an ortholog of the C. elegans unc-76 gene, which is necessary for normal axonal bundling and elongation within axon bundles. Expression of this gene in C. elegans unc-76 mutants can restore to the mutants partial locomotion and axonal fasciculation, suggesting that it also functions in axonal outgrowth. The N-terminal half of the gene product is highly acidic. Alternatively spliced transcript variants encoding different isoforms of this protein have been described. [provided by RefSeq
Sequence: C-QNKQKEQRELMKKSpecifications
| FEZ1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.01-0.03 ug/mL) The optimal working dilution should be determined by the end user. | |
| FEZ1 | |
| FEZ1 | |
| A synthetic peptide corresponding to human FEZ1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize isoform 1 (NP_005094.1) only. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FEZ1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_005094.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9638 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only