missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Ferret IgA/IgG/IgM (H+L) Secondary Antibody, Rhodamine, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73388
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Ferret samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
| IgA/IgG/IgM (H+L) | |
| Polyclonal | |
| Rhodamine | |
| Goat | |
| Affinity Purified | |
| RUO | |
| This reagent is suitable for the detection of all Ferret immunoglobulin isotypes and chain combinations. | |
| Ferret | |
| IgG |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| LYOPH | |
| Flow Cytometry 1:1:10 - 1:1000, Immunocytochemistry/Immunofluorescence 1:1000 - 1:5000, Fluorophore-linked immunosorbent assay 1:10000 - 1:50000 | |
| Ferret IgG, IgA, IgM whole molecules | |
| 1 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction