Learn More
Goat anti-Ferret IgA/IgG/IgM (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-73396
Product Code. 18298524
  • £328.00 / 1mg
Description
IgA/IgG/IgM (H+L) Polyclonal specifically detects IgA/IgG/IgM (H+L) in Ferret samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgA/IgG/IgM (H+L)
Polyclonal
Biotin
Goat
Affinity Purified
RUO
This product is suitable for the detection of all Ferret immunoglobulin classes, isotypes and chain combinations.
Ferret
IgG
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
LYOPH
Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000
Ferret IgG, IgA and IgM whole molecules
1 mg
Secondary
Reconstitute with 1.0 ml deionized water (or equivalent).
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.