missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Feline IgG F(ab')2 Secondary Antibody, PE/Cy5.5, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72798PECY55
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Feline samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| PBS | |
| Cat IgG F(ab')2 fragment | |
| 500 μg | |
| Adaptive Immunity, Epitope Tags, Immunology | |
| Feline | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| PE-Cyanine5.5 | |
| Goat | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction