Learn More
FBXO44, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FBXO44.
Brand: Abnova PAB6462.100ug
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. It is also a member of the NFB42 (neural F Box 42kDa) family, similar to F-box only protein 2 and F-box only protein 6. Four alternatively spliced transcript variants encoding two distinct isoforms have been found for this gene. [provided by RefSeq]
Sequence: C-AALTPPEPPSAEPSpecifications
| FBXO44 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of FBXO44. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_149438 | |
| FBXO44 | |
| A synthetic peptide corresponding to human FBXO44. | |
| RUO | |
| 93611 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:8000) Western blot (2-6.0 ug/mL) The optimal working dilution should be determined by the end user. | |
| FBXO44 | |
| DKFZp781J0852/FBG3/FBX30/FBX6A/Fbx44/Fbxo6a/MGC14140 | |
| Goat | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only