Learn More
FBXL3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FBXL3.
Brand: Abnova PAB7099.100ug
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats and is localized in the nucleus. [provided by RefSeq]
Sequence: KRGGRDSDRNSSEE-CSpecifications
| FBXL3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| FBXL3 | |
| FBL3/FBL3A/FBXL3A | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 26224 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FBXL3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_0.36290.1 | |
| FBXL3 | |
| A synthetic peptide corresponding to human FBXL3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only