Learn More
FABP3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of FABP3.
Brand: Abnova PAB27682.100ug
Description
The intracellular fatty acid-binding proteins (FABPs) belongs to a multigene family. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Fatty acid-binding protein 3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is a candidate tumor suppressor gene for human breast cancer. [provided by RefSeq
Sequence: PTTIIEKNGDILTLKSpecifications
| FABP3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.01-0.03ug/ml) The optimal working dilution should be determined by the end user. | |
| FABP3 | |
| FABP11/H-FABP/MDGI/O-FABP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2170 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of FABP3. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_004093.1 | |
| FABP3 | |
| A synthetic peptide corresponding to internal region of human FABP3. | |
| 100 μg | |
| Primary | |
| Chicken, Human, Mouse, Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only