missing translation for 'onlineSavingsMsg'
Learn More

FABP2, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of FABP2.

Brand:  Abnova PAB7258.100ug

Product Code. 16080348

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The intracellular fatty acid-binding proteins (FABPs) belong to a multigene family with nearly twenty identified members. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Intestinal fatty acid-binding protein 2 gene contains four exons and is an abundant cytosolic protein in small intestine epithelial cells. This gene has a polymorphism at codon 54 that identified an alanine-encoding allele and a threonine-encoding allele. Thr-54 protein is associated with increased fat oxidation and insulin resistance. [provided by RefSeq

Sequence: C-EGVEAKRIFKKD
Specifications

Specifications

FABP2
Polyclonal
Unconjugated
ELISA (1:64000) Western blot (0.001 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-3 ug/mL) The optimal working dilution should be determined by the end user.
FABP2
FABPI/I-FABP/MGC133132
Goat
Antigen affinity purification
RUO
2169
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Immunohistochemistry (PFA fixed), Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of FABP2.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_000125.2
FABP2
A synthetic peptide corresponding to C-terminus of human FABP2.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only