missing translation for 'onlineSavingsMsg'
Learn More

EZH1, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of EZH1.

Brand:  Abnova PAB6289.100ug

Product Code. 16089547

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EZH1 is a component of a noncanonical Polycomb repressive complex-2 (PRC2) that mediates methylation of histone H3 (see MIM 602812) lys27 (H3K27) and functions in the maintenance of embryonic stem cell pluripotency and plasticity (Shen et al., 2008 [PubMed 19026780]).[supplied by OMIM

Sequence: EIPNPPTSKCITY
Specifications

Specifications

EZH1
Polyclonal
Unconjugated
ELISA (1:2000) Western blot (2-6 ug/mL) The optimal working dilution should be determined by the end user.
EZH1
KIAA0388
Goat
Antigen affinity purification
RUO
2145
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of EZH1.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_001982.2
EZH1
A synthetic peptide corresponding to human EZH1.
100 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only