missing translation for 'onlineSavingsMsg'
Learn More

EXOC7, Goat, Polyclonal Antibody, Abnova™

Goat polyclonal antibody raised against synthetic peptide of EXOC7.

Brand:  Abnova PAB7019.100ug

Product Code. 16080138

  • £432.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EXOC7 is a component of the exocyst, which is an evolutionarily conserved octameric protein complex essential for exocytosis. The exocyst targets secretory vesicles at specific domains of the plasma membrane for cell surface expansion and protein secretion (Zuo et al., 2006 [PubMed 17086175]).[supplied by OMIM

Sequence: C-DNIKNDPDKEYN
Specifications

Specifications

EXOC7
Polyclonal
Unconjugated
ELISA (1:16000) Western blot (0.5-1.5 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user.
EXOC7
2-5-3p/DKFZp686J04253/EX070/EXO70/EXOC1/Exo70p/FLJ40965/FLJ46415/YJL085W
Goat
Antigen affinity purification
RUO
23265
Human
Liquid
ELISA, Immunohistochemistry (PFA fixed), Western Blot
0.5 mg/mL
Goat polyclonal antibody raised against synthetic peptide of EXOC7.
In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide)
NP_001013861.1;NP_0560.34.2
EXOC7
A synthetic peptide corresponding to human EXOC7.
100 μg
Primary
This antibody is expected to recognize both reported isoforms (NP_001013861.1 and NP_0560.34.2).
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only