Learn More
ETFB Goat anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ETFB.
Brand: Abnova PAB19033.100ug
Description
This gene encodes electron-transfer-flavoprotein, beta polypeptide, which shuttles electrons between primary flavoprotein dehydrogenases involved in mitochondrial fatty acid and amino acid catabolism and the membrane-bound electron transfer flavoprotein ubiquinone oxidoreductase. The gene deficiencies have been implicated in type II glutaricaciduria. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq
Sequence: C-QVTLEGDKLKVERESpecifications
| ETFB | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(3-5 ug/mL) ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ETFB | |
| FP585/MADD | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 2109 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ETFB. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001976.1;NP_001014763.1 | |
| ETFB | |
| A synthetic peptide corresponding to amino acids 152-165 at internal region of human ETFB. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only