missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Equine IgG F(ab')2 Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-73524
This item is not returnable.
View return policy
Description
IgG F(ab')2 Polyclonal antibody specifically detects IgG F(ab')2 in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG F(ab')2 | |
| Polyclonal | |
| HRP | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Horse IgG F(ab')2 fragment | |
| 2 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| LYOPH | |
| Western Blot 1:1000 - 1:5000, ELISA 1:50000 - 1:150000, Immunohistochemistry 1:500 - 1:2500, Dot Blot | |
| Goat | |
| Affinity Purified | |
| RUO | |
| No reaction was observed against Horse IgG F(c). | |
| Equine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction