Learn More
ELF1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ELF1.
Brand: Abnova PAB6207.100ug
Description
This gene encodes an E26 transformation-specific related transcription factor. The encoded protein is primarily expressed in lymphoid cells and acts as both an enhancer and a repressor to regulate transcription of various genes. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: C-AMKQNELLEPNSFSpecifications
| ELF1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ELF1 | |
| ELF1 | |
| A synthetic peptide corresponding to human ELF1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ELF1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_758961.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1997 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only