missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DYDC1 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DYDC1.
Brand: Abnova PAB18640.100ug
This item is not returnable.
View return policy
Description
Sequence: C-RNEDILHSEEATLDSpecifications
| DYDC1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) The optimal working dilution should be determined by the end user. | |
| DYDC1 | |
| DPY30D1/FLJ43920/bA36D19.5 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 143241 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of DYDC1. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_620167.1 | |
| DYDC1 | |
| A synthetic peptide corresponding to amino acids at internal region of human DYDC1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction