Learn More
DPM2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DPM2.
Brand: Abnova PAB17035.100ug
Description
Dolichol-phosphate mannose (Dol-P-Man) serves as a donor of mannosyl residues on the lumenal side of the endoplasmic reticulum (ER). Lack of Dol-P-Man results in defective surface expression of GPI-anchored proteins. Dol-P-Man is synthesized from GDP-mannose and dolichol-phosphate on the cytosolic side of the ER by the enzyme dolichyl-phosphate mannosyltransferase. The protein encoded by this gene is a hydrophobic protein that contains 2 predicted transmembrane domains and a putative ER localization signal near the C terminus. This protein associates with DPM1 in vivo and is required for the ER localization and stable expression of DPM1 and also enhances the binding of dolichol-phosphate to DPM1. [provided by RefSeq
Sequence: C-SYVMLKTKRVTKKAQSpecifications
| DPM2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) The optimal working dilution should be determined by the end user. | |
| DPM2 | |
| FLJ80013/MGC111193/MGC21559 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8818 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of DPM2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_003854.1 | |
| DPM2 | |
| A synthetic peptide corresponding to human DPM2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only