Learn More
DLL1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DLL1.
Brand: Abnova PAB12816.100ug
Description
DLL1 is a human homolog of the Notch Delta ligand and is a member of the delta/serrate/jagged family. It plays a role in mediating cell fate decisions during hematopoiesis. It may play a role in cell-to-cell communication. [provided by RefSeq
Sequence: C-ATQRHLTVGEEWSQDSpecifications
| DLL1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| DLL1 | |
| DELTA1/DL1/Delta | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 28514 | |
| Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of DLL1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_005609.3 | |
| DLL1 | |
| A synthetic peptide corresponding to amino acids at internal region of human DLL1. | |
| 100 μg | |
| Primary | |
| 75 KDa band observed in Mouse Heart and Rat Heart lysates (calculated MW of 78.1 KDa according to human NP_005609.3, 78.5K Da according to Mouse NP_031891.2 and 77.4 KDa according to Rat NP_114452.1). Recommended concentration: 0.3-1 ug/ml. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only