missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX19A/DDX19B, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DDX19A/DDX19B.
Brand: Abnova PAB7319.100ug
This item is not returnable.
View return policy
Description
Sequence: C-DLPVDKDGNPDNETYSpecifications
| DDX19A/DDX19B | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (0.5-1.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| NP_009173.1 (Gene ID : 11269);NP_060802.1 (Gene ID : 55308);NP_001014451.1 (Gene ID : 11269);NP_001014451.1 (Gene ID : 11269);NP_001014449.1 (Gene ID : 11269) | |
| DDX19A | |
| A synthetic peptide corresponding to human DDX19A/DDX19B. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize DDX19A (NP_060802.1) and all reported isoforms of DDX19B (NP_009173.1, NP_001014451.1 and NP_001014449.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of DDX19A/DDX19B. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| DDX19-DDX19L/DDX19L/DKFZp686C21137/FLJ11126 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 55308/11269 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction