Learn More
DCLRE1C, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DCLRE1C.
Brand: Abnova PAB10023.100ug
Description
This gene encodes a nuclear protein that is involved in V(D)J recombination and DNA repair. The protein has single-strand-specific 5'-3' exonuclease activity; it also exhibits endonuclease activity on 5' and 3' overhangs and hairpins when complexed with protein kinase, DNA-activated, catalytic polypeptide. Mutations in this gene cause Athabascan-type severe combined immunodeficiency (SCIDA). [provided by RefSeq]
Specifications
| DCLRE1C | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of DCLRE1C. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Q96SD1;NP_071932 | |
| DCLRE1C | |
| A synthetic peptide corresponding to amino acids 482-495 of human DCLRE1C. | |
| RUO | |
| 64421 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:10000-1:20000 Western Blot (1:500-1:2000) Immunohistochemistry (1:1000) The optimal working dilution should be determined by the end user. | |
| DCLRE1C | |
| A-SCID/DCLREC1C/FLJ11360/FLJ36438/RS-SCID/SCIDA/SNM1C | |
| Goat | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only