Learn More
DAG1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DAG1.
Brand: Abnova PAB18550.100ug
Description
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants
Sequence: C-HVGKHEYFMHATDKSpecifications
| DAG1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.2-0.6 ug/mL) The optimal working dilution should be determined by the end user. | |
| DAG1 | |
| 156DAG/A3a/AGRNR/DAG | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1605 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of DAG1. | |
| In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_004384.3 | |
| DAG1 | |
| A synthetic peptide corresponding to amino acids at internal region of human DAG1. | |
| 100 μg | |
| Primary | |
| Reported variants represent identical protein: NP_001159400.1, NP_004384.3. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only