missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4I1/COX4I2 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of COX4I1/COX4I2.
Brand: Abnova PAB7322.100ug
This item is not returnable.
View return policy
Description
Sequence: C-QGLASKWDYEKNESpecifications
| COX4I1/COX4I2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:4000) Western blot (0.01-0.03 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| NP_001852.1 | |
| COX4I1 | |
| A synthetic peptide corresponding to human COX4I1/COX4I2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of COX4I1/COX4I2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| COX4/COXIV/MGC72016 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1327/84701 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction