Learn More
COPS3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of COPS3.
Brand: Abnova PAB19717.100ug
Descrizione
The protein encoded by this gene possesses kinase activity that phosphorylates regulators involved in signal transduction. It phosphorylates I kappa-Balpha, p105, and c-Jun. It acts as a docking site for complex-mediated phosphorylation. The gene is located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Sequence: C-SLYKKNIQRLTSpecifica
| COPS3 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of COPS3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_003644.2 | |
| COPS3 | |
| A synthetic peptide corresponding to internal region of human COPS3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:4000) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| COPS3 | |
| CSN3/SGN3 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8533 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only