Learn More
COPA, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of COPA.
Brand: Abnova PAB27525.100ug
Description
In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by nonclathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms. [provided by RefSeq
Sequence: CTRASNLENSTSpecifications
| COPA | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (0.5-2ug/ml) The optimal working dilution should be determined by the end user. | |
| COPA | |
| FLJ26320/HEP-COP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1314 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of COPA. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_001091868.1;NP_004362.2 | |
| COPA | |
| A synthetic peptide corresponding to internal region of human COPA. | |
| 100 μg | |
| Primary | |
| Chicken, Human, Mouse, Pig, Rat |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only