Learn More
COMT Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of COMT.
Brand: Abnova PAB7078.100ug
Description
Catechol-O-methyltransferase catalyzes the transfer of a methyl group from S-adenosylmethionine to catecholamines, including the neurotransmitters dopamine, epinephrine, and norepinephrine. This O-methylation results in one of the major degradative pathways of the catecholamine transmitters. In addition to its role in the metabolism of endogenous substances, COMT is important in the metabolism of catechol drugs used in the treatment of hypertension, asthma, and Parkinson disease. COMT is found in two forms in tissues, a soluble form (S-COMT) and a membrane-bound form (MB-COMT). The differences between S-COMT and MB-COMT reside within the N-termini. Several transcript variants are formed through the use of alternative translation initiation sites and promoters. [provided by RefSeq
Sequence: C-QDIIPQLKKKYDVDSpecifications
| COMT | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Sandwich ELISA (0.5-2 ug/mL) Western Blot (0.03-0.1 ug/mL) The optimal working dilution should be determined by the end user. | |
| COMT | |
| COMT | |
| A synthetic peptide corresponding to internal region of human COMT. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_000745.1 and NP_009294.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of COMT. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000745.1;NP_009294.1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1312 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only