missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Goat anti-Chicken IgY (H+L) Secondary Antibody, FITC
Goat Polyclonal Secondary Antibody
Brand: Invitrogen™ PA128794
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Chicken secondary antibodies are affinity-purified antibodies with well-characterized specificity for the chicken immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
| Chicken IgY (H+L) | |
| Polyclonal | |
| FITC | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| Chicken | |
| Secondary Antibody | |
| IgG |
| ELISA, Flow Cytometry, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA and 0.01% sodium azide; pH 7.2 | |
| Chicken IgY whole molecule. | |
| 100 μg | |
| Secondary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction