missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Chicken IgM Heavy Chain Secondary Antibody, PE, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60690PE
This item is not returnable.
View return policy
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page
Specifications
| IgM Heavy Chain | |
| Polyclonal | |
| PBS | |
| Goat | |
| Multi-step | |
| RUO | |
| Secondary | |
| Chicken | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, SDS-Page | |
| PE | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Chicken IgM whole molecule | |
| 0.5 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction