missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Chicken IgM Heavy Chain Secondary Antibody, DyLight 594, Novus Biologicals™
Shop All Bio Techne ProductsDescription
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page
Specifications
Specifications
| Antigen | IgM Heavy Chain |
| Applications | Western Blot, ELISA, Immunohistochemistry, SDS-Page |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain |
| Host Species | Goat |
| Immunogen | Chicken IgM whole molecule |
| Purification Method | Multi-step |
| Quantity | 0.5 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?