Learn More
CDK10, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CDK10.
Brand: Abnova PAB13668.100ug
Description
The protein encoded by this gene belongs to the CDK subfamily of the Ser/Thr protein kinase family. The CDK subfamily members are highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2, and are known to be essential for cell cycle progression. This kinase has been shown to play a role in cellular proliferation and its function is limited to cell cycle G2-M phase. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: AEPDLECEQIRLKSpecifications
| CDK10 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CDK10 | |
| PISSLRE | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8558 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CDK10. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_443714.3 | |
| CDK10 | |
| A synthetic peptide corresponding to human CDK10. | |
| 100 μg | |
| Primary | |
| Approximately 37 KDa band observed in human lung lysates and in lysates of cell lines HEK293, A-549 and HepG2 (calculated MW of 41.0 KDa according to NP_443714.3). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only