Learn More
CDH11 Goat anti-Human, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CDH11.
Brand: Abnova PAB19043.100ug
Description
This gene encodes a type II classical cadherin from the cadherin superfamily, integral membrane proteins that mediate calcium-dependent cell-cell adhesion. Mature cadherin proteins are composed of a large N-terminal extracellular domain, a single membrane-spanning domain, and a small, highly conserved C-terminal cytoplasmic domain. Type II (atypical) cadherins are defined based on their lack of a HAV cell adhesion recognition sequence specific to type I cadherins. Expression of this particular cadherin in osteoblastic cell lines, and its upregulation during differentiation, suggests a specific function in bone development and maintenance. [provided by RefSeq
Sequence: CESDQTKPLSNQPSpecifications
| CDH11 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.1-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CDH11 | |
| CAD11/CDHOB/OB/OSF-4 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1009 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CDH11. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001788.2 | |
| CDH11 | |
| A synthetic peptide corresponding to amino acids 497-509 at internal region of human CDH11. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only