Learn More
CCBL1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CCBL1.
Brand: Abnova PAB7430.100ug
Description
This gene encodes a cytosolic enzyme that is responsible for the metabolism of cysteine conjugates of certain halogenated alkenes and alkanes. This metabolism can form reactive metabolites leading to nephrotoxicity and neurotoxicity. Increased levels of this enzyme have been linked to schizophrenia. Multiple transcript variants that encode different isoforms have been identified for this gene. [provided by RefSeq
Sequence: C-DISDFKRKMPDSpecifications
| CCBL1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) The optimal working dilution should be determined by the end user. | |
| CCBL1 | |
| FLJ95217/GTK/KAT1/KATI/MGC29624 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 883 | |
| Human | |
| Liquid |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CCBL1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_004050.3;NP_001116144.1 | |
| CCBL1 | |
| A synthetic peptide corresponding to human CCBL1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_004050.3, NP_001116144.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only